Novus Biologicals
Manufacturer Code:NBP246008
Catalog # NBP246008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3A2 |
Immunogen | Full length human recombinant protein of human HSPD1 (NP_955472) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 60 kDa chaperonin; 60 kDa chaperonin Chaperonin 60 CPN60 GROEL heat shock 60kD protein 1 (chaperonin) heat shock 60kDa protein 1 (chaperonin) Heat shock protein 60 heat shock protein 65 HLD4 Hsp60 HSP-60 HSP60SPG13 HSP65 HuCHA60 Mitochondrial matrix protein P160 kDa heat shock protein mitochondrial P60 lymphocyte protein short heat shock protein 60 Hsp60s1 spastic paraplegia 13 (autosomal dominant); 60 kDa heat shock protein, mitochondrial; Chaperonin 60; CPN60; heat shock 60kDa protein 1 (chaperonin); Heat shock protein 60; heat shock protein 65; HSP-60; HuCHA60; Mitochondrial matrix protein P1; P60 lymphocyte protein; short heat shock protein 60 Hsp60s1
Gene Aliases: CPN60; GROEL; HLD4; HSP-60; HSP60; HSP65; HSPD1; HuCHA60; SPG13
UniProt ID: (Human) P10809
Entrez Gene ID: (Human) 3329
Molecular Function:
chaperone
chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.