Novus Biologicals
Manufacturer Code:NBP122416IR
Catalog # NBP122416IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A7.H6 |
Immunogen | Full length protein HSP40 (YDJ1) |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DnaJ (Hsp40) homolog subfamily B member 1 DnaJ (Hsp40) homolog subfmaily B member 1 dnaJ homolog subfamily B member 1 DnaJ protein homolog 1 DNAJ1 hDj-1 HDJ1 Heat shock 40 kDa protein 1 heat shock 40kD protein 1 Heat shock protein 40 HSP40 HSPF1Hdj1 Human DnaJ protein 1 radial spoke 16 homolog B RSPH16B Sis1; DnaJ (Hsp40) homolog, subfamily B, member 1; DnaJ homolog subfamily B member 1; DnaJ protein homolog 1; hDj-1; Heat shock 40 kDa protein 1; HSP40; Human DnaJ protein 1; radial spoke 16 homolog B
Gene Aliases: DNAJ1; DNAJB1; HDJ1; Hsp40; HSPF1; RSPH16B; Sis1
UniProt ID: (Human) P25685
Entrez Gene ID: (Human) 3337
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.