Proteintech
Manufacturer Code:182841AP150UL
Catalog # 50-173-2090
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HSP27 Fusion Protein Ag13161 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28 kDa heat shock protein; epididymis secretory protein Li 102; Estrogen-regulated 24 kDa protein; Growth-related 25 kDa protein; Heat shock 25 kDa protein; Heat shock 27 kDa protein; heat shock 27kD protein 1; heat shock 27kDa protein 1; Heat shock protein beta 1 HSP 27 Hsp25 HSP27 HSP28 HSPB1; Heat shock protein beta-1; heat shock protein, 25 kDa; HSP 25; HSP 27; HspB1; p25; SRP27; Stress-responsive protein 27; truncated hsp25
Gene Aliases: 27kDa; CMT2F; HEL-S-102; HMN2B; HS.76067; Hsp25; HSP27; HSP28; HSPB1; SRP27
UniProt ID: (Human) P04792, (Mouse) P14602
Entrez Gene ID: (Human) 3315, (Mouse) 15507, (Rat) 24471
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.