Invitrogen
Manufacturer Code:MA538121
Catalog # PIMA538121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1092 |
Immunogen | A synthesized peptide derived from human Hsp105 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 105kDa 42 degrees C-HSP AI790491 Antigen NY-CO-25 heat shock 105/110 protein 1 heat shock 105kD alpha heat shock 105kD beta heat shock 105kDa heat shock 105kDa/110kDa protein 1 Heat shock 110 kDa protein Heat shock protein Heat shock protein 105 kDa heat shock protein 110 heat shock protein family H (Hsp110) member 1 heat shock protein 105 kDa heat shock-related 100 kDa protein E7I heat-shock protein 110 kDa HSP HSP105 HSP105 42 C-HSP HSP105A HSP105B Hsp110 hsp110/105 HSP-E7I HSPH1 I79_002075 KIAA0201 NY CO 25 NY-CO-25; 42 degrees C-HSP; Antigen NY-CO-25; heat shock 105kD alpha; heat shock 105kD beta; heat shock 105kDa; heat shock 105kDa/110kDa protein 1; Heat shock 110 kDa protein; Heat shock protein 105 kDa; heat shock protein 110; heat shock protein, 105 kDa; Heat shock-related 100 kDa protein E7I; HSP-E7I; HSP105 42 C-HSP
Gene Aliases: 105kDa; AI790491; hsp-E7I; HSP105; HSP105A; HSP105B; HSP110; hsp110/105; HSPH1; KIAA0201; NY-CO-25
UniProt ID: (Human) Q92598, (Mouse) Q61699, (Rat) Q66HA8
Entrez Gene ID: (Human) 10808, (Mouse) 15505, (Rat) 288444
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.