Invitrogen
Manufacturer Code:PA121213
Catalog # PIPA121213
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide: SGRKPQKTTSPT Synthetic peptide: NSQEEAEAKDEV and Synthetic peptide: FGEHYKRNETGL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: hormone - sensitive lipase testicular isoform; Hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; lipase E; lipase, hormone-sensitive; Monoacylglycerol lipase LIPE; REH; Retinyl ester hydrolase
Gene Aliases: 4933403G17Rik; AOMS4; FPLD6; HSL; LHS; LIPE
UniProt ID: (Human) Q05469, (Rat) P15304, (Mouse) P54310
Entrez Gene ID: (Human) 3991, (Rat) 25330, (Mouse) 16890
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.