Invitrogen
Manufacturer Code:PA19573
Catalog # 01-674-449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide based on mouse HSL protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: hormone - sensitive lipase testicular isoform; Hormone-sensitive lipase; HSL; HSL antibody hormone sensitive lipase antibody.; lipase E; Monoacylglycerol lipase LIPE; REH; Retinyl ester hydrolase
Gene Aliases: 4933403G17Rik; HSL; Lipe
UniProt ID: (Rat) P15304, (Mouse) P54310
Entrez Gene ID: (Rat) 25330, (Mouse) 16890
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.