Novus Biologicals
Manufacturer Code:NBP171931
Catalog # NBP171931
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 479 and 529 of human Heat Shock Transcription Factor 1 using the numbering given in entry NP_005517.1 (GeneID 3297). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Heat shock factor protein 1; heat shock factor protein 1 heat shock transcription factor 1HSTF1HSF 1 HSTF 1; Heat shock transcription factor 1; HSF 1; HSTF 1
Gene Aliases: HSF1; HSTF1
UniProt ID: (Human) Q00613
Entrez Gene ID: (Human) 3297
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.