Invitrogen
Manufacturer Code:PA5100564
Catalog # PIPA5100564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HSDL2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610207I16Rik C9orf99 HSDL2 hydroxysteroid dehydrogenase like 2 hydroxysteroid dehydrogenase-like protein 2 RGD1305387 SDR13C1 Short chain dehydrogenase/reductase family 13C member 1 short chain dehydrogenase/reductase family 13C member 1; Hydroxysteroid dehydrogenase-like protein 2; Short chain dehydrogenase/reductase family 13C member 1; short chain dehydrogenase/reductase family 13C, member 1
Gene Aliases: 2610207I16Rik; C9orf99; HSDL2; RGD1305387; SDR13C1
UniProt ID: (Human) Q6YN16, (Mouse) Q2TPA8, (Rat) Q4V8F9
Entrez Gene ID: (Human) 84263, (Mouse) 72479, (Rat) 313200
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.