Novus Biologicals
Manufacturer Code:NBP297749AF488
Catalog # NB103251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human HSDL2 fragment. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome 9 open reading frame 99 EC 1.1.1 EC 1.1.1.100 FLJ25855 hydroxysteroid dehydrogenase like 2 member 1 MGC10940; Hydroxysteroid dehydrogenase-like protein 2; Short chain dehydrogenase/reductase family 13C member 1; short chain dehydrogenase/reductase family 13C, member 1
Gene Aliases: C9orf99; HSDL2; SDR13C1
UniProt ID: (Human) Q6YN16
Entrez Gene ID: (Human) 84263
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.