Invitrogen
Manufacturer Code:PA575598
Catalog # PIPA575598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human HSD3B2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3 beta-HSD type II; 3 beta-hydroxysteroid dehydrogenase 2; 3 beta-hydroxysteroid dehydrogenase 6; 3 beta-hydroxysteroid dehydrogenase type II, delta 5-delta 4-isomerase type II, 3 beta-HSD type II; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 2; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 4; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type II; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type IV; 3-beta-HSD adrenal and gonadal type; 3-beta-HSD II; 3-beta-HSD IV; 3-beta-hydroxy-5-ene steroid dehydrogenase; 3-beta-hydroxy-Delta(5)-steroid dehydrogenase; 3beta-HSD HSD-3beta 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 2 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type II 3-beta-HSD II 3-beta-HSD adrenal and gonadal type 3-beta-hydroxy-Delta(5)-steroid dehydrogenase 3-beta-hydroxy-5-ene steroid dehydrogenaseProgesterone reductase Steroid Delta-isomerase Delta-5-3-ketosteroid isomerase HSD3B2 HSDB3B; delta 5-delta 4-isomerase type II; Delta-5-3-ketosteroid isomerase; Hydroxy-delta-5-steroid dehydrogenase 3 beta- and steroid delta-isomerase; hydroxysteroid dehydrogenase-2, delta<5>-3-beta; hydroxysteroid dehydrogenase-6 delta<5>-3-beta; Progesterone reductase; short chain dehydrogenase/reductase family 11E, member 2; Steroid Delta-isomerase
Gene Aliases: HSD3B; Hsd3b1; HSD3B2; Hsd3b6; HSDB; HSDB3B; SDR11E2
UniProt ID: (Human) P26439, (Mouse) P26149, (Rat) Q62878
Entrez Gene ID: (Human) 3284, (Mouse) 15493, (Rat) 29632
Molecular Function: dehydratase epimerase/racemase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.