Invitrogen
Manufacturer Code:PA5103519
Catalog # PIPA5103519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HSD3B1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3 beta-hydroxysteroid dehydrogenase 1; 3 beta-hydroxysteroid dehydrogenase 1 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I 3BETAHSD 3-beta-HSD 3-beta-HSD I 3-beta-hydroxy-5-ene steroid dehydrogenase 3-beta-hydroxy-Delta(5)-steroid dehydrogenase 3-beta-hydroxysteroid 3-dehydrogenase 3BH Delta-5-3-ketosteroid isomerase Dihydrotestosterone oxidoreductase HGNC:5217 HSD3B Hsd3b1 HSDB3 HSDB3A hydroxy-delta-5-steroid dehydrogenase 3 beta- and steroid delta-isomerase 1 hydroxy-delta-MAY-steroid dehydrogenase 3 beta- and steroid delta-isomerase hydroxysteroid dehydrogenase-1 delta<5>-3-beta I progesterone reductase SDR11E1 short chain dehydrogenase/reductase family 11E member 1 steroid Delta-isomerase Trophoblast antigen FDO161G unnamed protein product; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I; 3-beta-HSD I; 3-beta-hydroxy-5-ene steroid dehydrogenase; 3-beta-hydroxy-Delta(5)-steroid dehydrogenase; 3-beta-hydroxysteroid 3-dehydrogenase; Delta-5-3-ketosteroid isomerase; Dihydrotestosterone oxidoreductase; hydroxysteroid dehydrogenase-1, delta<5>-3-beta; progesterone reductase; short chain dehydrogenase/reductase family 11E, member 1; Steroid Delta-isomerase; Trophoblast antigen FDO161G
Gene Aliases: 3BETAHSD; 3BH; HSD3B; HSD3B1; HSDB3; HSDB3A; I; SDR11E1
UniProt ID: (Human) P14060, (Rat) P22071
Entrez Gene ID: (Human) 3283, (Rat) 360348
Molecular Function:
dehydratase
epimerase/racemase
isomerase
lyase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.