Novus Biologicals
Manufacturer Code:NBP233386
Catalog # NBP233386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: RHIVTLSNSVFTFSYKKAQRDLAYKPLYSWEEAKQKT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I 3BETAHSD 3-beta-HSD I 3-beta-hydroxy-Delta(5)-steroid dehydrogenase 3BH delta-5-3-ketosteroid isomerase EC 1.1.1 HSD3B HSDB3 HSDB3A hydroxy-delta-5-steroid dehydrogenase 3 beta- and steroid delta-isomerase 1 hydroxy-delta-5-steroid dehydrogenase 3 beta-and steroid delta-isomerase 1 I progesterone reductase SDR11E1 short chain dehydrogenase/reductase family 11E member 13-beta-hydroxy-5-ene steroid dehydrogenase steroid Delta-isomerase Trophoblast antigen FDO161G; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I; 3-beta-HSD I; 3-beta-hydroxy-5-ene steroid dehydrogenase; 3-beta-hydroxy-Delta(5)-steroid dehydrogenase; 3-beta-hydroxysteroid 3-dehydrogenase; Delta-5-3-ketosteroid isomerase; Dihydrotestosterone oxidoreductase; progesterone reductase; short chain dehydrogenase/reductase family 11E, member 1; Steroid Delta-isomerase; Trophoblast antigen FDO161G
Gene Aliases: 3BETAHSD; 3BH; HSD3B; HSD3B1; HSDB3; HSDB3A; I; SDR11E1
UniProt ID: (Human) P14060
Entrez Gene ID: (Human) 3283
Molecular Function: dehydratase epimerase/racemase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.