Invitrogen
Manufacturer Code:MA526466
Catalog # PIMA526466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6H2 |
Immunogen | Human recombit protein fragment corresponding to amino acids 57-261 of HSD17B8 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 8; 17-beta-hydroxysteroid dehydrogenase 8; 3-ketoacyl-[acyl-carrier-protein] reductase alpha subunit; 3-oxoacyl-[acyl-carrier-protein] reductase; D6S2245E FABG FABGL H2-KE6 HKE6 KE6 RING2 SDR30C1 dJ1033B10.9 17-beta-HSD 8 17-beta-hydroxysteroid dehydrogenase 8 3-oxoacyl-&lsqbacyl-carrier-protein] reductase estradiol 17-beta-dehydrogenase 8 estrogen 17-oxidoreductase ke-6 protein Ke6 really interesting new gene 2 protein short chain dehydrogenase/reductase family; Estradiol 17-beta-dehydrogenase 8; estrogen 17-oxidoreductase; hydroxysteroid (17-beta) dehydrogenase 8; KAR alpha subunit; Ke-6; Protein Ke6; Really interesting new gene 2 protein; Short chain dehydrogenase/reductase family 30C member 1; short chain dehydrogenase/reductase family 30C, member 1; Testosterone 17-beta-dehydrogenase 8
Gene Aliases: D6S2245E; dJ1033B10.9; FABG; FABGL; H2-KE6; HKE6; HSD17B8; KE6; RING2; SDR30C1
UniProt ID: (Human) Q92506
Entrez Gene ID: (Human) 7923, (Dog) 607895
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.