Novus Biologicals
Manufacturer Code:NBP271350AF350
Catalog # NP27130A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E12 |
Immunogen | Recombinant protein fragment corresponding to amino acids 57-261 of human HSD17B8 (NP_055049) produced in E.coli. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 17-beta-HSD 8; 17-beta-hydroxysteroid dehydrogenase 8; 3-ketoacyl-[acyl-carrier-protein] reductase alpha subunit; 3-oxoacyl-[acyl-carrier-protein] reductase; D6S2245EdJ1033B10.9 EC 1.1.1.- EC 1.1.1.62 EC 1.1.1.6317-beta-HSD 8 estrogen 17-oxidoreductase FABGLestradiol 17-beta-dehydrogenase 8 H2-KE6 HKE6FabG (beta-ketoacyl-[acyl-carrier-protein] reductase E coli) like (E. coli) hydroxysteroid (17-beta) dehydrogenase 83-oxoacyl-[acyl-carrier-protein] reductase ke-6 KE6 Ke-617-beta-hydroxysteroid dehydrogenase 8 Protein Ke6 Really interesting new gene 2 protein RING2FABG SDR30C1 short chain dehydrogenase/reductase family 30C member 1 Testosterone 17-beta-dehydrogenase 8; Estradiol 17-beta-dehydrogenase 8; estrogen 17-oxidoreductase; hydroxysteroid (17-beta) dehydrogenase 8; KAR alpha subunit; Ke-6; Protein Ke6; Really interesting new gene 2 protein; Short chain dehydrogenase/reductase family 30C member 1; short chain dehydrogenase/reductase family 30C, member 1; Testosterone 17-beta-dehydrogenase 8
Gene Aliases: D6S2245E; dJ1033B10.9; FABG; FABGL; H2-KE6; HKE6; HSD17B8; KE6; RING2; SDR30C1
UniProt ID: (Human) Q92506
Entrez Gene ID: (Human) 7923
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.