Proteintech
Manufacturer Code:148541AP150UL
Catalog # 50-172-9779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HSD17B7 Fusion Protein Ag6639 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17 beta HSD 7 3 keto steroid reductase HSD17B7 PRAP SDR37C1; 17 beta-hydroxysteroid dehydrogenase type VII; 17-beta-HSD 7; 17-beta-hydroxysteroid dehydrogenase 7; 17beta hydroxysteroid dehydrogenase; 3-keto-steroid reductase; 3-keto-steroid reductase/17-beta-hydroxysteroid dehydrogenase 7; Dihydrotestosterone oxidoreductase; Estradiol 17-beta-dehydrogenase 7; hydroxysteroid (17-beta) dehydrogenase 7; hydroxysteroid dehydrogenase 17 beta, type 7; PRAP; PRL receptor-associated protein; Short chain dehydrogenase/reductase family 37C member 1; short chain dehydrogenase/reductase family 37C, member 1
Gene Aliases: 17HSD7; AI266814; ERG27; HSD17B7; PRAP; SDR37C1; UNQ2563/PRO6243
UniProt ID: (Human) P56937, (Rat) Q62904, (Mouse) O88736
Entrez Gene ID: (Human) 51478, (Rat) 29540, (Mouse) 15490
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.