Novus Biologicals
Manufacturer Code:NBP299211F
Catalog # NB059054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human HSD17B6. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 6; 17-beta-HSD 6 17-beta-HSD6 17-beta-hydroxysteroid dehydrogenase type 6 3(alpha->beta)-hydroxysteroid epimerase 3-alpha->beta-HSE EC 1.1.1 EC 1.1.1.105 EC 1.1.1.62 EC 1.1.1.63 HSE3-hydroxysteroid epimerase hydroxysteroid (17-beta) dehydrogenase 6 hydroxysteroid (17-beta) dehydrogenase 6 homolog (mouse) NAD+ -dependent 3 alpha-hydroxysteroid dehydrogenase 3-hydroxysteroid epimerase Oxidative 3-alpha hydroxysteroid dehydrogenase oxidative 3-alpha-hydroxysteroid-dehydrogenase oxidoreductase retinol dehydrogenase RODH3(alpha->beta)-hydroxysteroid epimerasel SDR9C6 short chain dehydrogenase/reductase family 9C member 63-alpha->beta-hydroxysteroid epimerase; 17-beta-hydroxysteroid dehydrogenase type 6; 3(alpha->beta)-hydroxysteroid epimerase; 3(alpha->beta)-hydroxysteroid epimerasel; 3-alpha->beta-HSE; 3-alpha->beta-hydroxysteroid epimerase; 3-hydroxysteroid epimerase; hydroxysteroid (17-beta) dehydrogenase 6 homolog; NAD+ -dependent 3 alpha-hydroxysteroid dehydrogenase 3-hydroxysteroid epimerase; Oxidative 3-alpha hydroxysteroid dehydrogenase; oxidative 3-alpha-hydroxysteroid-dehydrogenase; oxidoreductase; retinol dehydrogenase; Short chain dehydrogenase/reductase family 9C member 6; short chain dehydrogenase/reductase family 9C, member 6
Gene Aliases: HSD17B6; HSE; RODH; SDR9C6
UniProt ID: (Human) O14756
Entrez Gene ID: (Human) 8630
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.