Invitrogen
Manufacturer Code:PA522367
Catalog # PIPA522367
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 18 and 261 of HSD17B4 (Uniprot IDNo.P51659) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (3R)-hydroxyacyl-CoA dehydrogenase; 17-beta-HSD 4; 17-beta-HSD IV; 17-beta-hydroxysteroid dehydrogenase 4; 17beta-estradiol dehydrogenase type IV; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase; beta-hydroxyacyl dehydrogenase; beta-keto-reductase; D-3-hydroxyacyl-CoA dehydratase; D-bifunctional protein; D-bifunctional protein, peroxisomal; DBP; DBP MFE-2 MPF-2 SDR8C1 17-beta-HSD 4 17-beta-HSD IV 17-beta-hydroxysteroid dehydrogenase 4 17beta-estradiol dehydrogenase type IV 3-alpha7-alpha12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase D-3-hydroxyacyl-CoA dehydratase D-bifunctional protein peroxisomal beta-hydroxyacyl dehydrogenase beta-keto-reductase multifunctional protein 2 peroxisomal multifunctional enzyme type 2 peroxisomal multifunctional protein 2 short chain dehydrogenase/reductase family 8C member 1; Enoyl-CoA hydratase 2; hydroxysteroid (17-beta) dehydrogenase 4; hydroxysteroid dehydrogenase 4; MFE-2; MFP-2; Multifunctional protein 2; Peroxisomal multifunctional enzyme type 2; peroxisomal multifunctional protein 2; Short chain dehydrogenase/reductase family 8C member 1; short chain dehydrogenase/reductase family 8C, member 1
Gene Aliases: 17-beta-HSD; 17[b]-HSD; DBP; 4; EDH17B4; HSD17B4; MFE-2; Mfp-2; MFP2; MPF-2; perMFE-2; PRLTS1; SDR8C1
UniProt ID: (Human) B4DVS5, (Mouse) P51660
Entrez Gene ID: (Human) 3295, (Mouse) 15488
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.