Invitrogen
Manufacturer Code:PA526524
Catalog # PIPA526524
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 89-118 amino acids from the central region of human HSD17B3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 3; 17-beta-HSD3; 17-beta-hydroxysteroid dehydrogenase type 3; EDH17B3 SDR12C2 17-beta-HSD 3 17-beta-HSD3 17-beta-hydroxysteroid dehydrogenase type 3 short chain dehydrogenase/reductase family 12C member 2 testicular 17-beta-hydroxysteroid dehydrogenase testosterone 17-beta-dehydrogenase 3; Estradiol 17-beta-dehydrogenase 2; hydroxysteroid (17-beta) dehydrogenase 3; hydroxysteroid dehydrogenase 3; Short chain dehydrogenase/reductase family 12C member 2; short chain dehydrogenase/reductase family 12C, member 2; Testicular 17-beta-hydroxysteroid dehydrogenase; Testosterone 17-beta-dehydrogenase 3
Gene Aliases: EDH17B3; HSD17B3; SDR12C2
UniProt ID: (Human) P37058
Entrez Gene ID: (Human) 3293
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.