Invitrogen
Manufacturer Code:MA525930
Catalog # PIMA525930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4F10 |
Immunogen | Full length human recombit protein of HSD17B2 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 17 beta HSD 217-beta-HSD 217-beta-hydroxysteroid dehydrogenase type 220 alpha HSD20 alpha hydroxysteroid dehydrogenase20 alpha-hydroxysteroid dehydrogenase20-alpha-HSDDHB2&lowbarHUMANE2DHEDH17B2Estradiol 17-beta-dehydrogenase 2HSD17Hsd17b2Hydroxysteroid (17 beta) dehydrogenase 2Microsomal 17 beta hydroxysteroid dehydrogenaseMicrosomal 17-beta-hydroxysteroid dehydrogenaseSDR9C2Testosterone 17 beta dehydrogenaseTestosterone 17-beta-dehydrogenase; 17-beta-HSD 2; 17-beta-hydroxysteroid dehydrogenase type 2; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E2DH; Estradiol 17-beta-dehydrogenase 2; hydroxysteroid (17-beta) dehydrogenase 2; Microsomal 17-beta-hydroxysteroid dehydrogenase; Short chain dehydrogenase/reductase family 9C member 2; short chain dehydrogenase/reductase family 9C, member 2; Testosterone 17-beta-dehydrogenase
Gene Aliases: EDH17B2; HSD17; HSD17B2; SDR9C2
UniProt ID: (Human) P37059
Entrez Gene ID: (Human) 3294
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.