Invitrogen
Manufacturer Code:PA544634
Catalog # PIPA544634
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human HSD17B14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 14; 17-beta-hydroxysteroid dehydrogenase 14; 17-beta-hydroxysteroid dehydrogenase DHRS10; dehydrogenase/reductase (SDR family) member 10; Dehydrogenase/reductase SDR family member 10; hydroxysteroid (17-beta) dehydrogenase 14; retinal short-chain dehydrogenase/reductase 3; Retinal short-chain dehydrogenase/reductase retSDR3; Short chain dehydrogenase/reductase family 47C member 1; short chain dehydrogenase/reductase family 47C, member 1; testicular tissue protein Li 52
Gene Aliases: DHRS10; HSD17B14; retSDR3; SDR3; SDR47C1; UNQ502/PRO474
UniProt ID: (Human) Q9BPX1
Entrez Gene ID: (Human) 51171
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.