Invitrogen
Manufacturer Code:PA525633
Catalog # PIPA525633
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 81-110 amino acids from the N-terminal region of human DHB13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta hydroxysteroid dehydrogenase; 17-beta hydroxysteroid dehydrogenase 13; 17-beta-HSD 13; 17-beta-hydroxysteroid dehydrogenase 13; Alcohol dehydrogenase PAN1B-like; hydroxysteroid (17-beta) dehydrogenase 13; NIIL497 SCDR9 SDR16C3 17-beta hydroxysteroid dehydrogenase 17-beta-HSD 13 17-beta-hydroxysteroid dehydrogenase 13 short chain dehydrogenase/reductase family 16C member 3 short-chain dehydrogenase/reductase 9; Short chain dehydrogenase/reductase family 16C member 3; short chain dehydrogenase/reductase family 16C, member 3; Short-chain dehydrogenase/reductase 9
Gene Aliases: AI047820; HMFN0376; HSD17B13; NIIL497; Pan1b; PAN1B-like; SCDR9; SDR16C3; UNQ497/PRO1014
UniProt ID: (Human) Q7Z5P4, (Mouse) Q8VCR2
Entrez Gene ID: (Human) 345275, (Mouse) 243168
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.