Novus Biologicals
Manufacturer Code:NBP304837100UL
Catalog # NB093426
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 20-300 of human HSD17B13 (NP_835236.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta hydroxysteroid dehydrogenase; 17-beta-HSD 13; 17-beta-hydroxysteroid dehydrogenase 13; HMFN0376 hydroxysteroid (17-beta) dehydrogenase 13 member 317-beta-hydroxysteroid dehydrogenase 13 MGC13850817-beta-HSD 13 NIIL497 SCDR9 short-chain dehydrogenase/reductase 9 UNQ497/PRO1014; hydroxysteroid (17-beta) dehydrogenase 13; Short chain dehydrogenase/reductase family 16C member 3; short chain dehydrogenase/reductase family 16C, member 3; Short-chain dehydrogenase/reductase 9
Gene Aliases: HMFN0376; HSD17B13; NIIL497; SCDR9; SDR16C3; UNQ497/PRO1014
UniProt ID: (Human) Q7Z5P4
Entrez Gene ID: (Human) 345275
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.