Abnova
Manufacturer Code:PAB22250
Catalog # 89-123-956
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human HSD17B13. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant HSD17B13.
For Research Use Only
Protein Aliases: 17-beta hydroxysteroid dehydrogenase; 17-beta-HSD 13; 17-beta-hydroxysteroid dehydrogenase 13; HMFN0376; hydroxysteroid (17-beta) dehydrogenase 13; MGC138508; MGC138510; NIIL497; SCDR9; SDR16C3; Short chain dehydrogenase/reductase family 16C member 3; short chain dehydrogenase/reductase family 16C, member 3; Short-chain dehydrogenase/reductase 9
Gene Aliases: HMFN0376; HSD17B13; NIIL497; SCDR9; SDR16C3; UNQ497/PRO1014
UniProt ID: (Human) Q7Z5P4
Entrez Gene ID: (Human) 345275
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.