Invitrogen
Manufacturer Code:PA569454
Catalog # PIPA569454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of mouse HSD17B12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 12; 17-beta-hydroxysteroid dehydrogenase 12; 17beta-HSD type 12; 3-ketoacyl-CoA reductase; Estradiol 17-beta-dehydrogenase 12; hydroxysteroid (17-beta) dehydrogenase 12; KAR; KAR SDR12C1 17-beta-HSD 12 17-beta-hydroxysteroid dehydrogenase 12 17beta-HSD type 12 3-ketoacyl-CoA reductase estradiol 17-beta-dehydrogenase 12 short chain dehydrogenase/reductase family 12C member 1 steroid dehydrogenase homolog; keratoadhesin; keratonectin; KIK-I; Short chain dehydrogenase/reductase family 12C member 1; short chain dehydrogenase/reductase family 12C, member 1; smooth muscle-specific 17 beta-hydroxysteroid dehydrogenase type 3; steroid dehydrogenase; steroid dehydrogenase homolog; Very-long-chain 3-oxoacyl-CoA reductase
Gene Aliases: 2610510O05Rik; AI172963; HSD17B12; KAR; KIK-I; Kik1; SDR12C1
UniProt ID: (Human) Q53GQ0, (Rat) Q6P7R8, (Mouse) O70503
Entrez Gene ID: (Pig) 100312972, (Human) 51144, (Rabbit) 100346603, (Dog) 475939, (Horse) 100050075, (Bovine) 789567, (Rat) 84013, (Mouse) 56348
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.