Invitrogen
Manufacturer Code:PA5112959
Catalog # PIPA5112959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Very-long-chain 3-oxoacyl-CoA reductase protein (40-181AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 12; 17-beta-HSD 12 17beta-HSD type 12 17-beta-hydroxysteroid dehydrogenase 12 2610510O05Rik 3-ketoacyl-CoA reductase AI172963 BOS_15148 Estradiol 17-beta-dehydrogenase 12 HSD17B12 hydroxysteroid (17-beta) dehydrogenase 12 hydroxysteroid 17-beta dehydrogenase 12 KAR keratoadhesin keratonectin Kik1 KIK-I LOC100346603 SDR12C1 Short chain dehydrogenase/reductase family 12C member 1 short chain dehydrogenase/reductase family 12C member 1 smooth muscle-specific 17 beta-hydroxysteroid dehydrogenase type 3 steroid dehydrogenase steroid dehydrogenase homolog Very-long-chain 3-oxoacyl-CoA reductase; 17-beta-hydroxysteroid dehydrogenase 12; 17beta-HSD type 12; 3-ketoacyl-CoA reductase; Estradiol 17-beta-dehydrogenase 12; hydroxysteroid (17-beta) dehydrogenase 12; KAR; Short chain dehydrogenase/reductase family 12C member 1; short chain dehydrogenase/reductase family 12C, member 1; steroid dehydrogenase homolog; Very-long-chain 3-oxoacyl-CoA reductase
Gene Aliases: HSD17B12; KAR; SDR12C1
UniProt ID: (Human) Q53GQ0
Entrez Gene ID: (Human) 51144
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.