Novus Biologicals
Manufacturer Code:NBP287605
Catalog # NB035227
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human HSD17B12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 12; 17-beta-HSD 12 17-beta-hydroxysteroid dehydrogenase 12 EC 1.1.1.623-ketoacyl-CoA reductase EC 1.3.1.- hydroxysteroid (17-beta) dehydrogenase 1217beta-HSD type 12 KARestradiol 17-beta-dehydrogenase 12 SDR12C1 short chain dehydrogenase/reductase family 12C member 1 steroid dehydrogenase homolog; 17-beta-hydroxysteroid dehydrogenase 12; 17beta-HSD type 12; 3-ketoacyl-CoA reductase; Estradiol 17-beta-dehydrogenase 12; hydroxysteroid (17-beta) dehydrogenase 12; KAR; Short chain dehydrogenase/reductase family 12C member 1; short chain dehydrogenase/reductase family 12C, member 1; steroid dehydrogenase homolog; Very-long-chain 3-oxoacyl-CoA reductase
Gene Aliases: HSD17B12; KAR; SDR12C1
UniProt ID: (Human) Q53GQ0
Entrez Gene ID: (Human) 51144
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.