Abnova
Manufacturer Code:H00003292M03A
Catalog # 89-104-687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2E5 |
Immunogen | HSD17B1 (NP_000404189 a.a. &bsim 285 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant HSD17B1.
For Research Use Only
Protein Aliases: 17-beta-HSD 1; 17-beta-hydroxysteroid dehydrogenase type 1; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E2DH; EDH17B2; EDHB17; Estradiol 17-beta-dehydrogenase 1; estradiol 17-beta-dehydrogenase-1; HSD17; hydroxysteroid (17-beta) dehydrogenase 1 isoform; MGC138140; Placental 17-beta-hydroxysteroid dehydrogenase; SDR28C1; Short chain dehydrogenase/reductase family 28C member 1; short chain dehydrogenase/reductase family 28CE, member 1
Gene Aliases: E17KSR; EDH17B1; EDH17B2; EDHB17; HSD17; HSD17B1; SDR28C1
UniProt ID: (Human) P14061
Entrez Gene ID: (Human) 3292
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.