Invitrogen
Manufacturer Code:PA542058
Catalog # PIPA542058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human HSD17B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-dehydrogenase 1; 17-beta-HSD 1; 17-beta-hydroxysteroid dehydrogenase type 1; 17beta-hydroxysteroid dehydrogenase type 1; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E2DH; Estradiol 17-beta-dehydrogenase 1; estradiol 17-beta-dehydrogenase-1; hydroxysteroid (17-beta) dehydrogenase 1 isoform; Hydroxysteroid dehydrogenase 17 beta, type 1; Placental 17-beta-hydroxysteroid dehydrogenase; Short chain dehydrogenase/reductase family 28C member 1; short chain dehydrogenase/reductase family 28CE, member 1
Gene Aliases: 17beta-HSD; 17BHD1; 17HSDB1; E17KSR; E2DH; EDH17B1; EDH17B2; EDHB17; HSD17; HSD17B1; Hsd17ba; SDR28C1
UniProt ID: (Human) P14061, (Mouse) P51656, (Rat) P51657
Entrez Gene ID: (Rabbit) 100009478, (Horse) 100036556, (Dog) 607570, (Bovine) 353107, (Pig) 100147712, (Human) 3292, (Mouse) 15485, (Guinea pig) 100718199, (Rat) 25322
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.