Invitrogen
Manufacturer Code:PA5115383
Catalog # PIPA5115383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HSD17B1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 1; 17-beta-hydroxysteroid dehydrogenase type 1; 17beta-HSD 17-beta-HSD 1 17-beta-hydroxysteroid dehydrogenase type 1 17BHD1 17HSDB1 20 alpha-hydroxysteroid dehydrogenase 20-alpha-HSD E17KSR E2DH EDH17B1 EDH17B2 EDHB17 Estradiol 17-beta-dehydrogenase 1 estradiol 17-beta-dehydrogenase-1 HSD17 HSD17B1 Hsd17ba hydroxysteroid (17-beta) dehydrogenase 1 hydroxysteroid (17-beta) dehydrogenase 1 isoform hydroxysteroid 17-beta dehydrogenase 1 Hydroxysteroid dehydrogenase 17 beta type 1 placental 17-beta-hydroxysteroid dehydrogenase SDR28C1 Short chain dehydrogenase/reductase family 28C member 1 short chain dehydrogenase/reductase family 28CE member 1; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E2DH; Estradiol 17-beta-dehydrogenase 1; estradiol 17-beta-dehydrogenase-1; hydroxysteroid (17-beta) dehydrogenase 1 isoform; Placental 17-beta-hydroxysteroid dehydrogenase; Short chain dehydrogenase/reductase family 28C member 1; short chain dehydrogenase/reductase family 28CE, member 1
Gene Aliases: E17KSR; EDH17B1; EDH17B2; EDHB17; HSD17; HSD17B1; SDR28C1
UniProt ID: (Human) P14061
Entrez Gene ID: (Human) 3292
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.