Invitrogen
Manufacturer Code:PA579399
Catalog # PIPA579399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human HSD11B2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: -HSD11 type II; 11(beta)-HSD2; 11-beta-HSD; 11-beta-HSD type II; 11-beta-HSD2; 11-beta-hydroxysteroid dehydrogenase type 2; 11-beta-hydroxysteroid dehydrogenase type II; 11-DH2; 11-HSD type II; Corticosteroid 11-beta-dehydrogenase isozyme 2; Corticosteroid 11-beta-dehydrogenase isozyme 2 11-beta-hydroxysteroid dehydrogenase type 2 11-DH2 11-beta-HSD2 11-beta-hydroxysteroid dehydrogenase type II 11-HSD type II 11-beta-HSD type II NAD-dependent 11-beta-hydroxysteroid dehydrogenase 11-beta-HSD Short chain dehydrogenase/reductase family 9C member 3 HSD11B2 HSD11K; hydroxysteroid (11-beta) dehydrogenase 2; Hydroxysteroid dehydrogenase, 11 beta type 2; NAD-dependent 11-beta-hydroxysteroid dehydrogenase; Short chain dehydrogenase/reductase family 9C member 3
Gene Aliases: 11HSD2; AME; AME1; HSD11B2; HSD11K; HSD2; SDR9C3
UniProt ID: (Human) P80365, (Mouse) P51661, (Rat) P50233
Entrez Gene ID: (Human) 3291, (Mouse) 15484, (Rat) 25117
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.