Invitrogen
Manufacturer Code:PA542758
Catalog # PIPA542758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human HSD11B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 11-beta-HSD1; 11-beta-hydroxysteroid dehydrogenase 1; 11-beta-hydroxysteroid dehydrogenase type 1; 11-DH; 11beta-HSD type 1; 11beta-HSD-1; 11beta-HSD1; 11beta-HSD1A; 11beta-hydroxysteroid dehydrogenase type 1; Corticosteroid 11-beta-dehydrogenase isozyme 1; Corticosteroid 11-beta-dehydrogenase, isozyme 1 (11-DH) (11-beta-hydroxysteroid dehydrogenase 1) (11-beta-HSD1) (11beta-HSD1A); cortocosteroid 11-beta dehydrogenase; hydroxysteroid (11-beta) dehydrogenase 1; Hydroxysteroid (11-beta) dehydrogenase 1 11-DH 11-beta-HSD1 HDL HSD11 HSD11B HSD11L MGC13539 SDR26C1; hydroxysteroid dehydrogenase, 11 beta type 1; Short chain dehydrogenase/reductase family 26C member 1; short chain dehydrogenase/reductase family 26C, member 1; short-chain reductase; short-chain reductase/dehydrogenase
Gene Aliases: 11-beta-HSD1; 11-DH; CORTRD2; HDL; HSD11; HSD11B; HSD11B1; HSD11L; LRRGT00065; SDR26C1
UniProt ID: (Human) P28845, (Sheep) P51975, (Guinea pig) Q6QLL4, (Rat) P16232, (Mouse) P50172
Entrez Gene ID: (Human) 3290, (Sheep) 443409, (Rabbit) 100342395, (Dog) 449023, (Bovine) 282589, (Horse) 100056429, (Guinea pig) 100135542, (Rat) 25116, (Mouse) 15483
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.