Proteintech
Manufacturer Code:109281AP150UL
Catalog # 50-172-6721
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HSD11B1 Fusion Protein Ag1361 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 11 beta HSD1 11 DH HDL HSD11 HSD11B HSD11B1 HSD11L SDR26C1; 11-beta-HSD1; 11-beta-hydroxysteroid dehydrogenase 1; 11-DH; 11beta-HSD-1; 11beta-HSD1A; 11beta-hydroxysteroid dehydrogenase type 1; Corticosteroid 11-beta-dehydrogenase isozyme 1; Corticosteroid 11-beta-dehydrogenase, isozyme 1 (11-DH) (11-beta-hydroxysteroid dehydrogenase 1) (11-beta-HSD1) (11beta-HSD1A); cortocosteroid 11-beta dehydrogenase; hydroxysteroid (11-beta) dehydrogenase 1; hydroxysteroid dehydrogenase, 11 beta type 1; Short chain dehydrogenase/reductase family 26C member 1; short chain dehydrogenase/reductase family 26C, member 1
Gene Aliases: 11-beta-HSD1; 11-DH; CORTRD2; HDL; HSD11; HSD11B; HSD11B1; HSD11L; LRRGT00065; SDR26C1
UniProt ID: (Human) P28845, (Rat) P16232, (Mouse) P50172
Entrez Gene ID: (Human) 3290, (Rat) 25116, (Mouse) 15483
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.