Invitrogen
Manufacturer Code:PA5116380
Catalog # PIPA5116380
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 528-562 amino acids from the C-terminal region of DANRE hspa8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410008N15Rik AAG2 constitutive heat shock protein 70 epididymis luminal protein 33 epididymis secretory sperm binding protein Li 72p FAM10A1 Heat shock 70 kDa protein 8 heat shock 70kd protein 10 heat shock 70kD protein 8 heat shock 70kDa protein 8 heat shock 70kDa protein 8-like heat shock cognate heat shock cognate 71 kDa protein heat shock cognate 71-kd protein heat shock cognate hsc73 heat shock cognate protein 54 Heat shock cognate protein 70 heat shock protein 8 heat shock protein A8 heat shock protein cognate 70 heat shock protein family A (Hsp70) member 8 HEL-33 HEL-S-72p HIP HSC54 Hsc70 HSC70 protein HSC71 Hsc73 HSP70 HSP71 Hsp73 HSPA10 Hspa8 hspa8 protein I79_022974 LAP LAP1 LAP-1 lipopolysaccharide-associated protein 1 LOC100348873 LPS-associated protein 1 MGC131511 MGC29929 NIP71 N-myristoyltransferase inhibitor protein 71 Protein FAM10A1 SNC6 wu:fb01g06 wu:fi48b06; heat shock 70 kDa protein 8; heat shock 70kDa protein 8; heat shock cognate 71-kd protein
Gene Aliases: hsc70; wu:fb01g06; wu:fi48b06
Entrez Gene ID: (Zebrafish) 573376
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.