Invitrogen
Manufacturer Code:PA547372
Catalog # PIPA547372
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit human Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Gln179-Arg605 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC-1 heparan sulfate 6-O-sulfotransferase 2; Heparan-sulfate 6-O-sulfotransferase 2; HS6ST-2
Gene Aliases: HS6ST2; PSEC0092
UniProt ID: (Human) Q96MM7
Entrez Gene ID: (Human) 90161
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.