Invitrogen
Manufacturer Code:PA5112578
Catalog # PIPA5112578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Heparan-sulfate 6-O-sulfotransferase 1 protein (3-133AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6Ost1 heparan sulfate 6-O-sulfotransferase 1 Heparan-sulfate 6-O-sulfotransferase 1 heparan-sulfate 6-sulfotransferase HH15 HS6ST Hs6st1 HS6ST-1 mHS6ST-1; Heparan-sulfate 6-O-sulfotransferase 1; heparan-sulfate 6-sulfotransferase; HS6ST-1; mHS6ST-1
Gene Aliases: 6Ost1; HH15; HS6ST; HS6ST1
UniProt ID: (Human) O60243, (Mouse) Q9QYK5
Entrez Gene ID: (Human) 9394, (Mouse) 50785
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.