Abnova
Manufacturer Code:H00009394A01
Catalog # 89-106-072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 303 a.a. &bsim 401 a.a) partial recombinant protein with GST tag.,HS6ST1 (NP_004798 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant HS6ST1.
For Research Use Only
Protein Aliases: DKFZp547H098; FLJ25392; Heparan-sulfate 6-O-sulfotransferase 1; heparan-sulfate 6-sulfotransferase; HS6ST; HS6ST-1; MGC116899; MGC116901
Gene Aliases: HH15; HS6ST; HS6ST1
UniProt ID: (Human) O60243
Entrez Gene ID: (Human) 9394
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.