Invitrogen
Manufacturer Code:PA569538
Catalog # PIPA569538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human HS3ST6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-OST-6; heparan sulfate (glucosamine) 3-O-sulfotransferase 5; heparan sulfate (glucosamine) 3-O-sulfotransferase 6; heparan sulfate 3-O-sulfotransferase 6; Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 6; Heparan sulfate glucosamine 3-O-sulfotransferase 6; heparan sulphate D-glucosaminyl 3-O-sulfotransferase-3B like; HS3ST6 HS3ST5
Gene Aliases: 3-OST-6; HS3ST5; HS3ST6
UniProt ID: (Human) Q96QI5, (Mouse) Q5GFD5
Entrez Gene ID: (Horse) 100146640, (Rabbit) 100346602, (Dog) 111096478, (Human) 64711, (Guinea pig) 100730401, (Rat) 684979, (Mouse) 328779
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.