Invitrogen
Manufacturer Code:PA543288
Catalog # PIPA543288
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human HS3ST5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-OST-5; h3-OST-5; heparan sulfate (glucosamine) 3-O-sulfotransferase 5; Heparan sulfate (glucosamine) 3-O-sulfotransferase 5 3-OST-5 3OST5 HS3OST5 NBLA04021; heparan sulfate (glucosamine) 3-O-sulfotransferase 6; Heparan sulfate 3-O-sulfotransferase 5; heparan sulfate 3-OST-5; Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 5; Heparan sulfate glucosamine 3-O-sulfotransferase 5
Gene Aliases: 3-OST-5; 3OST5; D930005L05Rik; Gm1151; HS3OST5; HS3ST5; NBLA04021
UniProt ID: (Human) Q8IZT8, (Mouse) Q8BSL4
Entrez Gene ID: (Human) 222537, (Horse) 100066938, (Dog) 481968, (Bovine) 540355, (Rabbit) 100346980, (Guinea pig) 100715395, (Mouse) 319415, (Rat) 294449
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.