Invitrogen
Manufacturer Code:PA542909
Catalog # PIPA542909
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human HS3ST3B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-OST-3B; heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1; Heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1 30ST3B1 3OST3B1; heparan sulfate 3-O-sulfotransferase 3B1; Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3B1; Heparan sulfate glucosamine 3-O-sulfotransferase 3B1; heparan sulfate-glucosamine 3-sulfotransferase 3B1
Gene Aliases: 3-OST-3B; 3OST3B1; h3-OST-3B; HS3ST3B; HS3ST3B1
UniProt ID: (Human) Q9Y662
Entrez Gene ID: (Bovine) 617069, (Human) 9953, (Rabbit) 100352865, (Horse) 100063069, (Pig) 100737777, (Rat) 303218, (Guinea pig) 100724084
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.