Invitrogen
Manufacturer Code:PA566502
Catalog # PIPA566502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human HS3ST3A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-OST-3A; 30ST3A1 3OST3A1 3-OST-3A h3-OST-3A heparan sulfate 3-O-sulfotransferase 3A1 heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3A1 heparan sulfate glucosamine 3-O-sulfotransferase 3A1 heparin-glucosamine 3-O-sulfotransferase; heparan sulfate (glucosamine) 3-O-sulfotransferase 3A1; heparan sulfate 3-O-sulfotransferase 3A1; Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3A1; Heparan sulfate glucosamine 3-O-sulfotransferase 3A1; heparin-glucosamine 3-O-sulfotransferase
Gene Aliases: 3-OST-3A; 3OST3A1; HS3ST3A; HS3ST3A1; UNQ2551/PRO6180
UniProt ID: (Human) Q9Y663
Entrez Gene ID: (Human) 9955
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.