Invitrogen
Manufacturer Code:PA531086
Catalog # PIPA531086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 217 of Human HS3ST2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-OST-2; 30ST2 3OST2 3-OST-2 h3-OST-2 heparan sulfate 3-O-sulfotransferase 2 heparan sulfate D-glucosaminyl 3-O-sulfotransferase 2 heparan sulfate glucosamine 3-O-sulfotransferase 2 heparin-glucosamine 3-O-sulfotransferase; h3-OST-2; heparan sulfate (glucosamine) 3-O-sulfotransferase 2; heparan sulfate 3-O-sulfotransferase 2; Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 2; Heparan sulfate glucosamine 3-O-sulfotransferase 2; heparin-glucosamine 3-O-sulfotransferase
Gene Aliases: 30ST2; 3OST2; HS3ST2; UNQ2442/PRO5004
UniProt ID: (Human) Q9Y278
Entrez Gene ID: (Human) 9956
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.