Novus Biologicals
Manufacturer Code:NBP287602
Catalog # NB035224
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human HS3ST2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-OST-2; EC 2.8.2 EC 2.8.2.293-OST-2 h3-OST-2 heparan sulfate (glucosamine) 3-O-sulfotransferase 230ST23OST2heparan sulfate glucosamine 3-O-sulfotransferase 2 Heparan sulfate 3-O-sulfotransferase 2 Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 2 heparin-glucosamine 3-O-sulfotransferase; h3-OST-2; heparan sulfate (glucosamine) 3-O-sulfotransferase 2; heparan sulfate 3-O-sulfotransferase 2; Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 2; Heparan sulfate glucosamine 3-O-sulfotransferase 2; heparin-glucosamine 3-O-sulfotransferase
Gene Aliases: 30ST2; 3OST2; HS3ST2; UNQ2442/PRO5004
UniProt ID: (Human) Q9Y278
Entrez Gene ID: (Human) 9956
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.