Proteintech
Manufacturer Code:143581AP150UL
Catalog # 50-172-9395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HS3ST1 Fusion Protein Ag5719 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3 OST 1 3OST 3OST1 h3 OST 1 HS3ST1; 3-OST-1; h3-OST-1; heparan sulfate (glucosamine) 3-O-sulfotransferase 1; Heparan sulfate 3-O-sulfotransferase 1; Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 1; Heparan sulfate glucosamine 3-O-sulfotransferase 1; heparin-glucosamine 3-O-sulfotransferase
Gene Aliases: 3-Ost; 3OST; 3OST1; D5Wsu110e; HS3ST1; Hsg3ost
UniProt ID: (Human) O14792, (Rat) Q9ESG5, (Mouse) O35310
Entrez Gene ID: (Human) 9957, (Rat) 84406, (Mouse) 15476
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.