Proteintech
Manufacturer Code:129301AP150UL
Catalog # 50-172-8355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HRSP12 Fusion Protein Ag3628 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 14.5 kDa translational inhibitor protein; 2-iminobutanoate/2-iminopropanoate deaminase; heat responsive protein 12 HRSP12 P14.5 PSP Ribonuclease UK114 UK114 UK114 antigen homolog; Heat-responsive protein 12; hp14.5; L-PSP; Liver perchloric acid-soluble protein; perchloric acid soluble protein; perchloric acid-soluble protein; perchrolic acid soluble protein; Reactive intermediate imine deaminase A homolog; reactive intermediate/imine deaminase A homolog; ribonuclease UK114; rp14.5; Translation inhibitor L-PSP ribonuclease; translational inhibitor p14.5; translational inhibitor protein p14.5; UK114 antigen homolog
Gene Aliases: HRSP12; P14.5; PSP; PSP1; RIDA; UK114
UniProt ID: (Rat) P52759, (Human) P52758
Entrez Gene ID: (Rat) 65151, (Human) 10247
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.