Proteintech
Manufacturer Code:664901IG150UL
Catalog # 50-173-6913
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4B4H2 |
Immunogen | HRPT2 CDC73 Fusion Protein ag2959 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C1orf28 CDC73 HPT JT HRPT2 Hyperparathyroidism 2 protein Parafibromin; cell division cycle 73 Paf1/RNA polymerase II complex component-like protein; cell division cycle 73, Paf1/RNA polymerase II complex component, homolog; Cell division cycle protein 73 homolog; hyperparathyroidism 2 (with jaw tumor); hyperparathyroidism 2 homolog; Hyperparathyroidism 2 protein; Hyperparathyroidism 2 protein homolog; Paf1/RNA polymerase II complex component; Parafibromin; Vcell division cycle 73, Paf1/RNA polymerase II complex component, homolog
Gene Aliases: 8430414L16Rik; BC027756; C130030P16Rik; C1orf28; C81219; CDC73; FIHP; HPTJT; HRPT1; HRPT2; HYX
UniProt ID: (Human) Q6P1J9, (Mouse) Q8JZM7, (Rat) Q4V8C8
Entrez Gene ID: (Human) 79577, (Pig) 100526068, (Mouse) 214498, (Rat) 304832
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.