Invitrogen
Manufacturer Code:PA594949
Catalog # PIPA594949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human HRH3 different from the related rat sequence by three amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 97; G-protein coupled receptor 97; H3R; HH3R; Histamine H3 receptor; Histamine H3 receptor H3R HH3R G-protein coupled receptor 97 HRH3 GPCR97
Gene Aliases: AW049250; GPCR97; H3R; HH3R; HRH3
UniProt ID: (Human) Q9Y5N1, (Rat) Q9QYN8, (Mouse) P58406
Entrez Gene ID: (Human) 11255, (Rat) 85268, (Mouse) 99296
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.