Invitrogen
Manufacturer Code:PA533847
Catalog # PIPA533847
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human HRH3 / Histamine H3 Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 97; G-protein coupled receptor 97; H3R; Histamine H3 receptor; HRH3 G protein-coupled receptor 97 GPCR97 H3 histamine receptor H3R HH3R Histamine H3 receptor Histamine receptor H3 G-protein coupled receptor 97 Histamine 3 receptor
Gene Aliases: GPCR97; HH3R; HRH3
UniProt ID: (Human) Q9Y5N1
Entrez Gene ID: (Human) 11255
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.