Invitrogen
Manufacturer Code:PA511701
Catalog # PIPA511701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 414-442 amino acids from the C-terminal region of human HRH3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 97; G-protein coupled receptor 97; H3R; Histamine H3 receptor; Histamine H3 receptor HH3R G-protein coupled receptor 97 HRH3 GPCR97
Gene Aliases: GPCR97; HH3R; HRH3
UniProt ID: (Human) Q9Y5N1
Entrez Gene ID: (Human) 11255
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.