Invitrogen
Manufacturer Code:PA5106845
Catalog # PIPA5106845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HRH3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW049250 G protein-coupled receptor 97 GPCR97 G-protein coupled receptor 97 H3R HH3R Histamine H3 receptor histamine receptor histamine receptor H3 Hrh3; G protein-coupled receptor 97; G-protein coupled receptor 97; H3R; HH3R; Histamine H3 receptor
Gene Aliases: AW049250; GPCR97; H3R; HH3R; HRH3
UniProt ID: (Human) Q9Y5N1, (Rat) Q9QYN8, (Mouse) P58406
Entrez Gene ID: (Human) 11255, (Rat) 85268, (Mouse) 99296
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.