Invitrogen
Manufacturer Code:PA521907
Catalog # PIPA521907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1 and 45 of Histamine H2 Receptor (Uniprot IDNo.P25021) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gastric receptor 1; Gastric receptor I; H2R; H2R HH2R gastric receptor 1 gastric receptor I histamine H2 receptor; HH2R; Histamine H2 receptor
Gene Aliases: H2R; HRH2
UniProt ID: (Human) P25021
Entrez Gene ID: (Human) 3274
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.